Learn More
THORLABS INC Thorlabs Kinematic Grating Mount Adapter, 20-40 mm Grating, Ø1″ Mirror Mount Compatible
Catalog No. NC3985261
Encompass
The KGM40 is a kinematic grating mount adapter designed for gratings with a height ranging from 20 to 40 mm. It is intended for use with Ø1″ mirror mounts.
  • Kinematic design
  • Compatible with gratings of 20 to 40 mm height
  • Designed for use with Ø1″ mirror mounts
  • RoHS compliant
  • REACH compliant
Catalog No. NC3985261 Supplier THORLABS INC Supplier No. KGM40
The KGM40 is a kinematic grating mount adapter designed for gratings with a height ranging from 20 to 40 mm. It is intended for use with Ø1″ mirror mounts.
  • Kinematic design
  • Compatible with gratings of 20 to 40 mm height
  • Designed for use with Ø1″ mirror mounts
  • RoHS compliant
  • REACH compliant