Learn More
THORLABS INC Thorlabs Lens Mount, Retaining Ring, 1 in. Optics, 8-32 Tap
Catalog No. NC3985243
Encompass
This lens mount is designed for 1 in. optics and includes a retaining ring. It features an 8-32 tap for secure mounting and is available as a 5-pack.
  • Lens mount with retaining ring for 1 in. optics
  • 8-32 tap
  • Available as a 5-pack
  • RoHS compliant
  • REACH compliant
  • Lifetime warranty
Catalog No. NC3985243 Supplier THORLABS INC Supplier No. LMR1P5
This lens mount is designed for 1 in. optics and includes a retaining ring. It features an 8-32 tap for secure mounting and is available as a 5-pack.
  • Lens mount with retaining ring for 1 in. optics
  • 8-32 tap
  • Available as a 5-pack
  • RoHS compliant
  • REACH compliant
  • Lifetime warranty