Learn More
THORLABS INC THORLABS Mounted Cylindrical Lens, 200.0 mm Focal Length, 1 in. Diameter, N-BK7
Catalog No. NC3985326
Encompass
This Thorlabs mounted cylindrical lens features a 200.0 mm focal length and a 1 in. diameter. Constructed from N-BK7 glass, it is designed with an anti-reflection coating for wavelengths ranging from 650 nm to 1050 nm.
  • Mounted plano-convex round cylindrical lens
  • 200.0 mm focal length
  • 1 in. diameter
  • N-BK7 glass material
  • Anti-reflection coating for 650 nm to 1050 nm wavelength range
Catalog No. NC3985326 Supplier THORLABS INC Supplier No. LJ1653RMB
This Thorlabs mounted cylindrical lens features a 200.0 mm focal length and a 1 in. diameter. Constructed from N-BK7 glass, it is designed with an anti-reflection coating for wavelengths ranging from 650 nm to 1050 nm.
  • Mounted plano-convex round cylindrical lens
  • 200.0 mm focal length
  • 1 in. diameter
  • N-BK7 glass material
  • Anti-reflection coating for 650 nm to 1050 nm wavelength range