Learn More
THORLABS INC THORLABS Mounted Plano-Convex Cylindrical Lens, 50.0 mm, 1 in., N-BK7, ARC 350-700
Catalog No. NC3985321
Encompass
This mounted plano-convex round cylindrical lens features a 50.0 mm focal length and a 1 in. diameter. It is constructed from N-BK7 material and includes an ARC 350-700 coating.
  • Mounted plano-convex round cylindrical lens
  • N-BK7 material
  • ARC 350-700 coating
  • 50.0 mm focal length
  • 1 in. diameter
Catalog No. NC3985321 Supplier THORLABS INC Supplier No. LJ1695RMA
This mounted plano-convex round cylindrical lens features a 50.0 mm focal length and a 1 in. diameter. It is constructed from N-BK7 material and includes an ARC 350-700 coating.
  • Mounted plano-convex round cylindrical lens
  • N-BK7 material
  • ARC 350-700 coating
  • 50.0 mm focal length
  • 1 in. diameter