Learn More
THORLABS INC Thorlabs Mounted Plano-Convex Cylindrical Lens, N-BK7, f=100 mm, Ø1 in., ARC 350-700 nm
Catalog No. NC3985318
Encompass
This mounted plano-convex cylindrical lens is made of N-BK7. It has a focal length of 100.0 mm and a diameter of 1 in. The lens features an anti-reflective coating for the 350 to 700 nm wavelength range.
  • Focal length of 100.0 mm.
  • Diameter of 1 in.
  • Constructed from N-BK7 material.
  • Anti-reflective coating for 350 to 700 nm wavelength range.
  • RoHS compliant.
  • REACH compliant.
  • Two-year warranty.
Catalog No. NC3985318 Supplier THORLABS INC Supplier No. LJ1567RMA
This mounted plano-convex cylindrical lens is made of N-BK7. It has a focal length of 100.0 mm and a diameter of 1 in. The lens features an anti-reflective coating for the 350 to 700 nm wavelength range.
  • Focal length of 100.0 mm.
  • Diameter of 1 in.
  • Constructed from N-BK7 material.
  • Anti-reflective coating for 350 to 700 nm wavelength range.
  • RoHS compliant.
  • REACH compliant.
  • Two-year warranty.