Learn More
THORLABS INC Thorlabs Mounted Plano-Convex Round Cylindrical Lens, 50.0 mm Focal Length, 1 in. Diameter, N-BK7
Catalog No. NC3985333
Encompass
The Thorlabs LJ1695RM is a mounted plano-convex round cylindrical lens with a focal length of 50.0 mm and a diameter of 1 in. Made from N-BK7 material, this lens is designed for various optical applications.
  • Focal length of 50.0 mm.
  • Diameter of 1 in.
  • Constructed from N-BK7 material.
  • Compliant with RoHS regulations.
  • Compliant with REACH regulations.
  • Two year warranty.
Catalog No. NC3985333 Supplier THORLABS INC Supplier No. LJ1695RM
The Thorlabs LJ1695RM is a mounted plano-convex round cylindrical lens with a focal length of 50.0 mm and a diameter of 1 in. Made from N-BK7 material, this lens is designed for various optical applications.
  • Focal length of 50.0 mm.
  • Diameter of 1 in.
  • Constructed from N-BK7 material.
  • Compliant with RoHS regulations.
  • Compliant with REACH regulations.
  • Two year warranty.