Learn More
THORLABS INC THORLABS Powder-Free Latex Gloves, Medium, Latex, 100 Count
Catalog No. NC3985295
Encompass
These powder-free latex gloves are designed for optics handling, providing protection and preventing the transfer of oils and debris. They feature textured fingertips for improved grip and come in a convenient pack of 100.
  • Disposable design for single-use applications.
  • Prevents the transfer of oils and debris from hands to optics.
  • Features textured fingertips to aid with picking up optics.
  • Each pack contains 100 gloves for ample supply.
  • Medium size with a palm circumference of 7.5 in. to 8 in. (19.05 cm to 20.32 cm).
Catalog No. NC3985295 Supplier THORLABS INC Supplier No. MC12M
These powder-free latex gloves are designed for optics handling, providing protection and preventing the transfer of oils and debris. They feature textured fingertips for improved grip and come in a convenient pack of 100.
  • Disposable design for single-use applications.
  • Prevents the transfer of oils and debris from hands to optics.
  • Features textured fingertips to aid with picking up optics.
  • Each pack contains 100 gloves for ample supply.
  • Medium size with a palm circumference of 7.5 in. to 8 in. (19.05 cm to 20.32 cm).