Learn More
THORLABS INC Thorlabs Rotation Mount, Ø1″ Optics, 360° Continuous, 15° Indexed
Catalog No. NC3985245
Encompass
This rotation mount is designed for Ø1″ optics, offering both continuous 360° rotation and 15° indexed increments. It is ideal for polarization optics applications requiring precise alignment.
  • Directly mounts Ø1″ optics up to 0.50″ (12.7 mm) thick.
  • The engraved scale can be rotated independently of the mounted optic for alignment.
  • Allows switching between continuous and discrete rotation.
  • Provides discrete rotation in 15° increments.
  • Features a 5 arcmin Vernier scale, ideal for polarization optics.
  • Includes one retaining ring.
  • Post mountable via an 8-32 (M4) Tap.
  • Top-located setscrew allows switching between smooth continuous 360° motion and discrete incremental rotation.
  • Internal threading: 1.035″-40.
  • Adjustable zero.
Catalog No. NC3985245 Supplier THORLABS INC Supplier No. RSP1X15
This rotation mount is designed for Ø1″ optics, offering both continuous 360° rotation and 15° indexed increments. It is ideal for polarization optics applications requiring precise alignment.
  • Directly mounts Ø1″ optics up to 0.50″ (12.7 mm) thick.
  • The engraved scale can be rotated independently of the mounted optic for alignment.
  • Allows switching between continuous and discrete rotation.
  • Provides discrete rotation in 15° increments.
  • Features a 5 arcmin Vernier scale, ideal for polarization optics.
  • Includes one retaining ring.
  • Post mountable via an 8-32 (M4) Tap.
  • Top-located setscrew allows switching between smooth continuous 360° motion and discrete incremental rotation.
  • Internal threading: 1.035″-40.
  • Adjustable zero.