Learn More
THORLABS INC Thorlabs Unthreaded Mounting Bushing, 1 in. Clear Aperture, 1.5 in. Mounting Hole, Aluminum
Catalog No. NC3985279
Encompass
This unthreaded mounting bushing is designed for use with 38.1 mm (1.5 in.) posts to mount 25.4 mm (1 in.) optics. It features a durable aluminum construction with a black anodized finish, providing both stability and a knurled edge for easier handling.
  • 25.4 mm (1 in.) clear aperture
  • 38.1 mm (1.5 in.) mounting hole
  • Constructed from aluminum
  • Black anodized finish
  • Compatible with 38.1 mm (1.5 in.) posts
  • Features a knurled edge for easier handling
  • RoHS compliant
Catalog No. NC3985279 Supplier THORLABS INC Supplier No. MC10BL
This unthreaded mounting bushing is designed for use with 38.1 mm (1.5 in.) posts to mount 25.4 mm (1 in.) optics. It features a durable aluminum construction with a black anodized finish, providing both stability and a knurled edge for easier handling.
  • 25.4 mm (1 in.) clear aperture
  • 38.1 mm (1.5 in.) mounting hole
  • Constructed from aluminum
  • Black anodized finish
  • Compatible with 38.1 mm (1.5 in.) posts
  • Features a knurled edge for easier handling
  • RoHS compliant