Learn More
THORLABS INC THORLABS Zero-Order Vortex Half-Wave Plate, 1 in., m=1, 830 nm
Catalog No. NC3985334
Encompass
These zero-order, m = 1 vortex half-wave plates affect radial and azimuthal polarization. They have constant retardance across the clear aperture, with a continuously rotating fast axis.
  • When viewed through crossed polarizers with white light, they produce an intensity profile with 2 modulations.
  • With linearly polarized light, these retarders generate an m = 2 polarization pattern.
  • The donut hole intensity profile from m = 1 retarders is smaller and more circular than m = 2 retarders.
  • These polarization sensitive devices produce different output polarizations based on the wave plate's fast axis orientation to the input beam.
  • They are mounted in an aluminum housing with an engraving to assist in locating the plate's center point for beam alignment.
  • The zero-degree fast axis is indicated by 3 lines.
Catalog No. NC3985334 Supplier THORLABS INC Supplier No. WPV10L830
These zero-order, m = 1 vortex half-wave plates affect radial and azimuthal polarization. They have constant retardance across the clear aperture, with a continuously rotating fast axis.
  • When viewed through crossed polarizers with white light, they produce an intensity profile with 2 modulations.
  • With linearly polarized light, these retarders generate an m = 2 polarization pattern.
  • The donut hole intensity profile from m = 1 retarders is smaller and more circular than m = 2 retarders.
  • These polarization sensitive devices produce different output polarizations based on the wave plate's fast axis orientation to the input beam.
  • They are mounted in an aluminum housing with an engraving to assist in locating the plate's center point for beam alignment.
  • The zero-degree fast axis is indicated by 3 lines.