Learn More
ATCC THP-1, Acute Monocytic Leukemia, Frozen
Supplier:  ATCC TIB202/DEL
Encompass
These cells are suitable as a transfection host. The cells are phagocytic (for both latex beads and sensitized erythrocytes) and lack surface and cytoplasmic immunoglobulin. Monocytic differentiation can be induced with the phorbol ester 12-O-tetradecanoylphorbol-13-acetate (TPA). For use by FOR-PROFIT companies only; NON-PROFIT companies use 50188860NP. New ATCC customers are required to register with ATCC prior to purchase. The application can be found at https://www.atcc.org/en/My_Profile/Register.aspx
Catalog No. 50-188-860FP