Learn More
Medchemexpress LLC THP-PEG3-OH | 60221-37-6 | 98.7% | 1 G
SDP
Catalog No. 5000340054
Encompass_Preferred
THP-PEG3-OH is a PEG-based PROTAC linker used in the synthesis of PROTACs. This compound is intended for research use only and is not sold to patients.
  • Enables the synthesis of PROTACs
  • Functions as a PROTAC linker
  • Utilizes the PROTAC pathway
  • Facilitates selective degradation of target proteins via the ubiquitin-proteasome system
Catalog No. 50-003-40054 Supplier Medchemexpress LLC Supplier No. HY1302161G
THP-PEG3-OH is a PEG-based PROTAC linker used in the synthesis of PROTACs. This compound is intended for research use only and is not sold to patients.
  • Enables the synthesis of PROTACs
  • Functions as a PROTAC linker
  • Utilizes the PROTAC pathway
  • Facilitates selective degradation of target proteins via the ubiquitin-proteasome system