Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC THP-PEG4-OH | 128660-97-9 | ≥98.0% | 500 MG
SDP

Catalog No. 5000340493
Encompass_Preferred

THP-PEG4-OH is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs.

  • Molecular weight: 278.34
  • Formula: C13H26O6
  • Appearance: Liquid
  • Color: Colorless to light yellow
  • SMILES: OCCOCCOCCOCCOC1CCCCO1
  • Contains two different ligands connected by a linker for E3 ubiquitin ligase and target protein
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins

Catalog No. 50-003-40493 Supplier Medchemexpress LLC Supplier No. HY130230500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

THP-PEG4-OH is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs.

  • Molecular weight: 278.34
  • Formula: C13H26O6
  • Appearance: Liquid
  • Color: Colorless to light yellow
  • SMILES: OCCOCCOCCOCCOC1CCCCO1
  • Contains two different ligands connected by a linker for E3 ubiquitin ligase and target protein
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.