Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC THP-PEG4-Pyrrolidine(N-Boc)-CH2OH | 2378261-80-2 | 95.0% | 25 MG
SDP

Catalog No. 5000342773
Encompass_Preferred

THP-PEG4-Pyrrolidine(N-Boc)-CH2OH is a PEG-based PROTAC linker primarily used in the synthesis of PROTAC K-Ras Degrader-1. This compound serves as a crucial component for designing degraders that target specific proteins, utilizing the intracellular ubiquitin-proteasome system. It is intended for research use only.

  • PEG-based PROTAC linker
  • Used in the synthesis of PROTAC K-Ras degrader-1
  • Purity of 95.0%
  • Molecular weight is 477.59
  • Molecular formula is C23H43NO9
  • Appearance is a colorless to light yellow liquid
  • Store at -20°C under nitrogen
  • Soluble in DMSO

Catalog No. 50-003-42773 Supplier Medchemexpress LLC Supplier No. 25MGHY130820
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

THP-PEG4-Pyrrolidine(N-Boc)-CH2OH is a PEG-based PROTAC linker primarily used in the synthesis of PROTAC K-Ras Degrader-1. This compound serves as a crucial component for designing degraders that target specific proteins, utilizing the intracellular ubiquitin-proteasome system. It is intended for research use only.

  • PEG-based PROTAC linker
  • Used in the synthesis of PROTAC K-Ras degrader-1
  • Purity of 95.0%
  • Molecular weight is 477.59
  • Molecular formula is C23H43NO9
  • Appearance is a colorless to light yellow liquid
  • Store at -20°C under nitrogen
  • Soluble in DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.