Learn More
Medchemexpress LLC THP-PEG4-Pyrrolidine(N-Me)-CH2OH | 2378261-81-3 | 95.0% | 50 MG
SDP
Catalog No. 5000340278
Encompass_Preferred
THP-PEG4-Pyrrolidine(N-Me)-CH2OH is a PEG-based PROTAC linker. This compound is used in the synthesis of PROTACs, which are molecules designed to leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins. PROTACs achieve this by connecting two ligands: one for an E3 ubiquitin ligase and another for the target protein, offering a novel approach in biological activity modulation.
  • Functions as a PROTAC linker
  • Facilitates selective protein degradation
  • Useful for biological research applications
  • Available with comprehensive documentation
Catalog No. 50-003-40278 Supplier Medchemexpress LLC Supplier No. HY13082150MG
THP-PEG4-Pyrrolidine(N-Me)-CH2OH is a PEG-based PROTAC linker. This compound is used in the synthesis of PROTACs, which are molecules designed to leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins. PROTACs achieve this by connecting two ligands: one for an E3 ubiquitin ligase and another for the target protein, offering a novel approach in biological activity modulation.
  • Functions as a PROTAC linker
  • Facilitates selective protein degradation
  • Useful for biological research applications
  • Available with comprehensive documentation