Learn More
Invivogen THP1-Lucia NF-κB Cells, Engineered Monocytic Reporter Cells, Mycoplasma-Free, 3-7 x 10⁶ cells
SDP
Catalog No. NC2947887
Encompass
The THP1-Lucia NF-κB Cells are advanced engineered human THP-1 monocytic reporter cells. They allow sensitive monitoring of the NF-κB signaling pathway using secreted Lucia luciferase activity, offering a convenient method that eliminates cell lysis. This product stands out with an improved reporter/background signal ratio and responds effectively to various PRR agonists and cytokines. Ideal for studies in immunology, virology, and innate immune response.
Catalog No. NC2947887 Supplier Invivogen Supplier No. THPLNFKBV2
The THP1-Lucia NF-κB Cells are advanced engineered human THP-1 monocytic reporter cells. They allow sensitive monitoring of the NF-κB signaling pathway using secreted Lucia luciferase activity, offering a convenient method that eliminates cell lysis. This product stands out with an improved reporter/background signal ratio and responds effectively to various PRR agonists and cytokines. Ideal for studies in immunology, virology, and innate immune response.