Learn More
Medchemexpress LLC Thp104c | 877792-12-6 | 99.0% | 376.43 g/mol | C20H16N4O2S | 5 MG
SDP
Catalog No. 5000221633
Encompass_Preferred
THP104c is a small-molecule inhibitor of mitochondrial fission (CAS 877792-12-6) intended for research use. The compound has molecular formula C20H16N4O2S and a molecular weight of 376.43 g/mol. It is supplied as an off-white to light yellow solid with reported high purity, and documentation such as a data sheet, COA, and SDS is available to support lab use.
  • Inhibits mitochondrial fission for cell biology and mitochondrial research.
  • High reported purity suitable for in vitro assays and biochemical studies.
  • Off-white to light yellow solid for straightforward storage and handling.
  • Available in multiple laboratory-scale quantities to support experiments.
  • Includes COA and SDS for quality verification and safety information.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-21633 Supplier Medchemexpress LLC Supplier No. HY1449065MG
THP104c is a small-molecule inhibitor of mitochondrial fission (CAS 877792-12-6) intended for research use. The compound has molecular formula C20H16N4O2S and a molecular weight of 376.43 g/mol. It is supplied as an off-white to light yellow solid with reported high purity, and documentation such as a data sheet, COA, and SDS is available to support lab use.
  • Inhibits mitochondrial fission for cell biology and mitochondrial research.
  • High reported purity suitable for in vitro assays and biochemical studies.
  • Off-white to light yellow solid for straightforward storage and handling.
  • Available in multiple laboratory-scale quantities to support experiments.
  • Includes COA and SDS for quality verification and safety information.
  • For research use only; not for human or clinical use.