Learn More
Chem-Impex International, Inc. THPTA | MFCD27665386 | 250MG
Supplier:  Chem-Impex International, Inc. 35126250MG
Encompass_Preferred
THPTA 99 - 100% (HPLC) 250mg CAS#760952-88-3 MFCD27665386 C18H30N10O3 MW:434.50
Catalog No. NC1976075