Learn More
Bachem (Thr4,Gly7)-Oxytocin
SDP
Supplier:  Bachem 4013837.0005
Encompass_Preferred
(Thr4,Gly7)-Oxytocin , H-Cys-Tyr-Ile-Thr-Asn-Cys-Gly-Leu-Gly-NH2, 60786-59-6, C39H61N11O12S2, Mr 940.11, 5mg. TGOT. TGOT exhibits a 640-fold increase in oxytocic/ antidiuretic selectivity relative to oxytocin.
Catalog No. 50-260-164