Learn More
Aviva Systems Biology Corp THRAP5 Antibody - C-terminal region (ARP32301_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32301_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against THRAP5. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human THRAP5. Alternate names: DRIP92, THRAP5, TRAP95ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rat
Catalog No. 50-014-54518