Learn More
Aviva Systems Biology Corp Thrb Antibody - N-terminal region (ARP36994_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36994_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against Thrb. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of mouse Thrb. Alternate names: Nr1a2, T3R[b], T3Rbeta, Thrb1, Thrb2, c-erbAbetaApplicationCHIP, WB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-56305