Learn More
Aviva Systems Biology Corp THRB Antibody - middle region (ARP37995_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37995_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against THRB. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human THRB. Alternate names: ERBA-BETA, ERBA2, GRTH, MGC126109, MGC126110, NR1A2, THR1, THRB1, THRB2, PRTH, C-ERBA-2, C-ERBA-BETAApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-56481