Learn More
Grainger Smith-Cooper Threaded 45-Degree Forged Elbow, Carbon Steel, 2000 lb, 1/2 in., 1 ea.
Catalog No. 502787445
Encompass_Preferred
This threaded 45-degree forged elbow is designed for piping system connections requiring a directional change. Constructed from durable carbon steel, this fitting is engineered for use in demanding industrial applications.
  • Forged carbon steel construction
  • Nominal size of 1/2 in.
  • Pressure class rating of 2000 lb.
  • NPT end style for secure connection
  • Conforms to ASME B1.20.1 standards
Catalog No. 50-278-7445 Supplier Grainger Supplier No. 195V11
This threaded 45-degree forged elbow is designed for piping system connections requiring a directional change. Constructed from durable carbon steel, this fitting is engineered for use in demanding industrial applications.
  • Forged carbon steel construction
  • Nominal size of 1/2 in.
  • Pressure class rating of 2000 lb.
  • NPT end style for secure connection
  • Conforms to ASME B1.20.1 standards