Learn More
MSC E-Z LOK Thread Locking Insert, 3/8-16, 9/16-12, Carbon Steel, 10/Pk.
Catalog No. 502869935
Encompass
The E-Z LOK Thread Locking Insert is designed for thread repair, featuring a 3/8-16 internal thread and a 9/16-12 external thread. It adheres to the UNC thread standard and has an overall length of 0.515 in. The insert is made from carbon steel and has a bright/uncoated finish.
  • Internal thread size: 3/8-16
  • External thread size: 9/16-12
  • Insert length: 0.515 in.
  • Material: Carbon steel
  • Thread-locking insert type: Thread repair
  • Thread standard: UNC
  • Coating/finish: Bright/uncoated
Catalog No. 50-286-9935 Supplier MSC Supplier No. 80939903
The E-Z LOK Thread Locking Insert is designed for thread repair, featuring a 3/8-16 internal thread and a 9/16-12 external thread. It adheres to the UNC thread standard and has an overall length of 0.515 in. The insert is made from carbon steel and has a bright/uncoated finish.
  • Internal thread size: 3/8-16
  • External thread size: 9/16-12
  • Insert length: 0.515 in.
  • Material: Carbon steel
  • Thread-locking insert type: Thread repair
  • Thread standard: UNC
  • Coating/finish: Bright/uncoated