Learn More
Chemglass Life Sciences Chem-Thread, Size 18, 22mm Tube OD, Accepts 14-18mm OD Tubing, #208 O-ring
SDP
Catalog No. 501531629
Encompass_Preferred
Chem-Thread, Size 18, 22mm Tube OD, Accepts 14-18mm OD Tubing, #208 O-ring. Consists of an externally threaded glass tube, Viton o-ring and Celcon compression cap. Tube O.D. listed is the shank O.D. of the connector.
Catalog No. 50-153-1629 Supplier Chemglass Life Sciences Supplier No. CG35004
Chem-Thread, Size 18, 22mm Tube OD, Accepts 14-18mm OD Tubing, #208 O-ring. Consists of an externally threaded glass tube, Viton o-ring and Celcon compression cap. Tube O.D. listed is the shank O.D. of the connector.