Learn More
BURKLE INC Thread adapter S 55x5 (DIN51) inner - G2" inner, green
Catalog No. 5000025678
Encompass_Preferred
This adapter is used when pumps need to be attached to containers such as drums or canisters with different thread standards.It is also suitable for laboratories, where experimental setups often require creative connections, as our adapters provide a secure and adaptable interface.
Catalog No. 50-000-25678 Supplier BURKLE INC Supplier No. 06002005
This adapter is used when pumps need to be attached to containers such as drums or canisters with different thread standards.It is also suitable for laboratories, where experimental setups often require creative connections, as our adapters provide a secure and adaptable interface.