Learn More
BURKLE INC Thread adapter S64x5 outer- S61x6 (DIN61) inner, yellow/blue
Catalog No. 5000025679
Encompass_Preferred
This adapter is used when pumps need to be attached to containers such as drums or canisters with different thread standards.It is also suitable for laboratories, where experimental setups often require creative connections, as our adapters provide a secure and adaptable interface.
Catalog No. 50-000-25679 Supplier BURKLE INC Supplier No. 06001013
This adapter is used when pumps need to be attached to containers such as drums or canisters with different thread standards.It is also suitable for laboratories, where experimental setups often require creative connections, as our adapters provide a secure and adaptable interface.