Learn More
BURKLE INC Thread adapter ETFE GL45-GL32
Catalog No. 5000025951
Encompass_Preferred
The thread adapter was specially designed as an accessory for the Mini solvent pump. It allows great flexibility in the choice of container for the hand pump. Thanks to its robust ETFE/PTFE body and the GL-45 connection standard, it enables simple and safe connection of the solvent pump to smaller container openings.
Due to its high chemical resistance, the adapter is particularly suitable for use with aggressive media in demanding laboratory and industrial environments. The high-quality workmanship ensures a secure, tight fit.
Catalog No. 50-000-25951 Supplier BURKLE INC Supplier No. 00064003
The thread adapter was specially designed as an accessory for the Mini solvent pump. It allows great flexibility in the choice of container for the hand pump. Thanks to its robust ETFE/PTFE body and the GL-45 connection standard, it enables simple and safe connection of the solvent pump to smaller container openings.
Due to its high chemical resistance, the adapter is particularly suitable for use with aggressive media in demanding laboratory and industrial environments. The high-quality workmanship ensures a secure, tight fit.