Learn More
BURKLE INC Thread adapter S 61x6 (DIN61) outer- G 2" outer, black
Catalog No. 5000026023
Encompass_Preferred
This adapter is used when pumps need to be attached to containers such as drums or canisters with different thread standards.It is also suitable for laboratories, where experimental setups often require creative connections, as our adapters provide a secure and adaptable interface.
Catalog No. 50-000-26023 Supplier BURKLE INC Supplier No. 06003007
This adapter is used when pumps need to be attached to containers such as drums or canisters with different thread standards.It is also suitable for laboratories, where experimental setups often require creative connections, as our adapters provide a secure and adaptable interface.