Learn More
BURKLE INC Thread adapter S64x5 outer- S55x5 (DIN51) inner, black
Catalog No. 5000025537
Encompass_Preferred
This adapter is used when pumps need to be attached to containers such as drums or canisters with different thread standards.It is also suitable for laboratories, where experimental setups often require creative connections, as our adapters provide a secure and adaptable interface.
Catalog No. 50-000-25537 Supplier BURKLE INC Supplier No. 06001016
This adapter is used when pumps need to be attached to containers such as drums or canisters with different thread standards.It is also suitable for laboratories, where experimental setups often require creative connections, as our adapters provide a secure and adaptable interface.