Learn More
BURKLE INC Thread adapter S61x6 (DIN61) outer S56x4(T-S) outer,black
Catalog No. 5000025824
Encompass_Preferred
This adapter is used when pumps need to be attached to containers such as drums or canisters with different thread standards.It is also suitable for laboratories, where experimental setups often require creative connections, as our adapters provide a secure and adaptable interface.
Catalog No. 50-000-25824 Supplier BURKLE INC Supplier No. 06003006
This adapter is used when pumps need to be attached to containers such as drums or canisters with different thread standards.It is also suitable for laboratories, where experimental setups often require creative connections, as our adapters provide a secure and adaptable interface.