Learn More
BURKLE INC Thread adapter S70x6 (Mauser) outer- S64x5 outer,green
Catalog No. 5000026061
Encompass_Preferred
This adapter is used when pumps need to be attached to containers such as drums or canisters with different thread standards.It is also suitable for laboratories, where experimental setups often require creative connections, as our adapters provide a secure and adaptable interface.
Catalog No. 50-000-26061 Supplier BURKLE INC Supplier No. 06003004
This adapter is used when pumps need to be attached to containers such as drums or canisters with different thread standards.It is also suitable for laboratories, where experimental setups often require creative connections, as our adapters provide a secure and adaptable interface.