Learn More
Analytical Sales & Services Thread Adapter PP GL45 Cap - GL32 Bottle
Catalog No. 5000805416
Encompass
Thread Adapter PP GL45 Cap - GL32 Bottle for Canary-Safe Caps
Catalog No. 50-008-05416 Supplier Analytical Sales & Services Supplier No. CA4504
Thread Adapter PP GL45 Cap - GL32 Bottle for Canary-Safe Caps