Learn More
Grainger Thread Cutting Screw #10 Pan 5/8 L PK100
Catalog No. 502748901
Encompass_Preferred
The Approved Vendor Thread Cutting Screw is a self-tapping screw made of alloy steel with a black oxide finish. It has a pan head, type F point, and is available in sizes from #6-24 to 1/4"-20. The screws are designed for use in thin metal, plastic, fiberglass and wood. They create their own thread pattern when driven into a pre-drilled pilot hole and provide a strong, reusable threaded fastener. The black oxide coating offers mild corrosion resistance. Sold in packs of 25, 50 or 100.
Catalog No. 50-274-8901 Supplier Grainger Supplier No. 14N249
The Approved Vendor Thread Cutting Screw is a self-tapping screw made of alloy steel with a black oxide finish. It has a pan head, type F point, and is available in sizes from #6-24 to 1/4"-20. The screws are designed for use in thin metal, plastic, fiberglass and wood. They create their own thread pattern when driven into a pre-drilled pilot hole and provide a strong, reusable threaded fastener. The black oxide coating offers mild corrosion resistance. Sold in packs of 25, 50 or 100.