Learn More
Grainger Thread Mill 1/16 1/8 -27 Carbide
Catalog No. 502767566
Encompass_Preferred
The Micro 100 16R675 is a 1/4" diameter, 3/4" long, 4 flute carbide thread mill with a right hand spiral flute. It has a TiAlN coating and can cut 60-62 HRC materials. The thread mill has a 1/4"-28 UNF shank and can cut thread sizes from #10-24 through 1/4"-28. It has a neck relief below the thread for improved chip clearance. The Micro 100 16R675 thread mill is made in the USA.
Catalog No. 50-276-7566 Supplier Grainger Supplier No. 16R675
The Micro 100 16R675 is a 1/4" diameter, 3/4" long, 4 flute carbide thread mill with a right hand spiral flute. It has a TiAlN coating and can cut 60-62 HRC materials. The thread mill has a 1/4"-28 UNF shank and can cut thread sizes from #10-24 through 1/4"-28. It has a neck relief below the thread for improved chip clearance. The Micro 100 16R675 thread mill is made in the USA.