Learn More
Grainger Thread Mill 1/2 -13 Carbide
Catalog No. 502766485
Encompass_Preferred
The Micro 100 16R708 is a carbide thread mill with a 1/4" shank diameter, 1/2" max cutting diameter, and 1-1/2" overall length. It has a 60 degree thread angle and can cut unified coarse threads from 4 to 56 TPI in steel, stainless steel, cast iron, aluminum, and other materials. The carbide construction provides wear resistance and tool life. This thread mill is made in the USA by Micro 100, a manufacturer of indexable carbide and solid carbide cutting tools.
Catalog No. 50-276-6485 Supplier Grainger Supplier No. 16R708
The Micro 100 16R708 is a carbide thread mill with a 1/4" shank diameter, 1/2" max cutting diameter, and 1-1/2" overall length. It has a 60 degree thread angle and can cut unified coarse threads from 4 to 56 TPI in steel, stainless steel, cast iron, aluminum, and other materials. The carbide construction provides wear resistance and tool life. This thread mill is made in the USA by Micro 100, a manufacturer of indexable carbide and solid carbide cutting tools.