Learn More
Grainger Thread Mill 1/2 3/4 -14 Carbide
Catalog No. 502765862
Encompass_Preferred
The Micro 100 16R667 is a 1/4" diameter, 3/4" long, 4 flute carbide thread mill with a TiAlN coating. It has a right hand spiral flute and can cut 60 degree internal threads in steel, stainless steel, cast iron, aluminum, and other materials. The thread mill has a 1/2" shank and is capable of cutting unified thread sizes from 4-56 to 1/4-20 as well as metric thread sizes from M3 to M6. With its sharp cutting edges and wear resistant coating, this thread mill provides accurate threading and long tool life in production applications.
Catalog No. 50-276-5862 Supplier Grainger Supplier No. 16R667
The Micro 100 16R667 is a 1/4" diameter, 3/4" long, 4 flute carbide thread mill with a TiAlN coating. It has a right hand spiral flute and can cut 60 degree internal threads in steel, stainless steel, cast iron, aluminum, and other materials. The thread mill has a 1/2" shank and is capable of cutting unified thread sizes from 4-56 to 1/4-20 as well as metric thread sizes from M3 to M6. With its sharp cutting edges and wear resistant coating, this thread mill provides accurate threading and long tool life in production applications.