Learn More
Grainger Thread Mill 1/2 3/4 -14 Carbide
Catalog No. 502766028
Encompass_Preferred
The Micro 100 16R664 is a 1/2" diameter, 4 flute, carbide thread mill with a TiAlN coating. It has a right hand spiral flute and can cut 60 degree threads. The overall length is 3" and it has a 1/2" shank. This thread mill is designed for milling internal threads in steel, stainless steel, cast iron, aluminum and other non-ferrous metals. It has a tolerance of +/- .0005" and can cut unified, metric, acme, buttress and other thread forms. The TiAlN coating provides heat and wear resistance. This is a high performance thread mill for demanding applications.
Catalog No. 50-276-6028 Supplier Grainger Supplier No. 16R664
The Micro 100 16R664 is a 1/2" diameter, 4 flute, carbide thread mill with a TiAlN coating. It has a right hand spiral flute and can cut 60 degree threads. The overall length is 3" and it has a 1/2" shank. This thread mill is designed for milling internal threads in steel, stainless steel, cast iron, aluminum and other non-ferrous metals. It has a tolerance of +/- .0005" and can cut unified, metric, acme, buttress and other thread forms. The TiAlN coating provides heat and wear resistance. This is a high performance thread mill for demanding applications.