Learn More
Grainger Thread Mill 1/4 3/8 -18 Carbide
Catalog No. 502768609
Encompass_Preferred
The Micro 100 16R671 is a 1/4" diameter carbide thread mill with a 60 degree thread angle. It has 4 flutes and can cut right hand threads from 4 to 56 threads per inch. The overall length is 3" and it has a 1/2" shank. This thread mill is made of solid carbide for wear resistance when cutting steel, stainless steel, cast iron, aluminum, and other materials. It can be run at speeds up to 2000 SFM.
Catalog No. 50-276-8609 Supplier Grainger Supplier No. 16R671
The Micro 100 16R671 is a 1/4" diameter carbide thread mill with a 60 degree thread angle. It has 4 flutes and can cut right hand threads from 4 to 56 threads per inch. The overall length is 3" and it has a 1/2" shank. This thread mill is made of solid carbide for wear resistance when cutting steel, stainless steel, cast iron, aluminum, and other materials. It can be run at speeds up to 2000 SFM.