Learn More
Grainger Thread Mill #1-8 Carbide
Supplier:  Grainger 16R677
Encompass_Preferred
The Micro 100 16R677 is a 1/4" diameter, 4 flute, carbide thread mill with a TiAlN coating. It has a right hand spiral flute and can cut 60 degree threads. The overall length is 2-1/2" and it has a 1/2" shank. This thread mill is designed for high speed machining in steels, stainless steels, cast iron, and non-ferrous materials. It can be run at speeds up to 2000 SFM. The TiAlN coating provides heat and wear resistance. This is a high performance thread mill for demanding applications.
Catalog No. 50-276-5992