Learn More
Grainger Thread Mill 3/4 -10 Carbide
Catalog No. 502765689
Encompass_Preferred
The Grainger 16R716 is a Westward 3/8-inch drive impact socket adapter that converts a 3/8-inch drive to a 1/2-inch drive. It is made from chrome vanadium steel for durability and has a black phosphate finish to resist corrosion. This impact socket adapter allows you to use 3/8-inch drive tools with a 1/2-inch drive impact wrench, increasing versatility. It measures 2-1/2 inches long and weighs 0.19 lbs. The 16R716 socket adapter is ideal for automotive repair shops, fleet maintenance, and other heavy duty applications that require frequent socket changes.
Catalog No. 50-276-5689 Supplier Grainger Supplier No. 16R716
The Grainger 16R716 is a Westward 3/8-inch drive impact socket adapter that converts a 3/8-inch drive to a 1/2-inch drive. It is made from chrome vanadium steel for durability and has a black phosphate finish to resist corrosion. This impact socket adapter allows you to use 3/8-inch drive tools with a 1/2-inch drive impact wrench, increasing versatility. It measures 2-1/2 inches long and weighs 0.19 lbs. The 16R716 socket adapter is ideal for automotive repair shops, fleet maintenance, and other heavy duty applications that require frequent socket changes.