Learn More
Grainger Thread Mill 3/4 -12 Carbide
Catalog No. 502768534
Encompass_Preferred
The Thread Mill 3/4-12 Carbide is a high-performance thread milling cutter made by Niagara Cutter. It has a 3/4" shank diameter and can cut 12 threads per inch. The cutter is made of solid carbide for durability and has TiAlN coating for heat and wear resistance. It can machine a wide range of materials including steel, stainless steel, cast iron, aluminum, titanium and high temp alloys. The 3-flute design provides efficient chip evacuation. It is capable of high feed rates and productivity in thread milling applications. The cutter produces excellent thread quality and dimensional accuracy.
Catalog No. 50-276-8534 Supplier Grainger Supplier No. 16R717
The Thread Mill 3/4-12 Carbide is a high-performance thread milling cutter made by Niagara Cutter. It has a 3/4" shank diameter and can cut 12 threads per inch. The cutter is made of solid carbide for durability and has TiAlN coating for heat and wear resistance. It can machine a wide range of materials including steel, stainless steel, cast iron, aluminum, titanium and high temp alloys. The 3-flute design provides efficient chip evacuation. It is capable of high feed rates and productivity in thread milling applications. The cutter produces excellent thread quality and dimensional accuracy.