Learn More
Grainger Thread Mill 3/8 -24 Carbide
Catalog No. 502765687
Encompass_Preferred
The Grainger 16R702 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is totally enclosed and fan cooled with Class F insulation and a C-face endshield for easy mounting. It can provide 0.56 kW of power with 1.8 nominal amps at full load. This electric motor is designed for general purpose applications.
Catalog No. 50-276-5687 Supplier Grainger Supplier No. 16R702
The Grainger 16R702 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is totally enclosed and fan cooled with Class F insulation and a C-face endshield for easy mounting. It can provide 0.56 kW of power with 1.8 nominal amps at full load. This electric motor is designed for general purpose applications.