Learn More
Grainger Thread Mill 5/16 -18 Carbide
Catalog No. 502767575
Encompass_Preferred
The Grainger 16R695 is a Westward 3/8-inch drive deep impact socket set. This 15-piece set includes sizes ranging from 4mm to 14mm as well as a 3-inch extension. The sockets are made from chrome vanadium steel for durability and have a black phosphate finish to resist corrosion. The set comes packed in a molded case for storage and portability. With deep broached hexagons, these impact sockets are designed to grip fasteners tightly for torque and longevity. Overall, this is a quality impact socket set for automotive and industrial use.
Catalog No. 50-276-7575 Supplier Grainger Supplier No. 16R695
Add to Cart
Add to Cart
The Grainger 16R695 is a Westward 3/8-inch drive deep impact socket set. This 15-piece set includes sizes ranging from 4mm to 14mm as well as a 3-inch extension. The sockets are made from chrome vanadium steel for durability and have a black phosphate finish to resist corrosion. The set comes packed in a molded case for storage and portability. With deep broached hexagons, these impact sockets are designed to grip fasteners tightly for torque and longevity. Overall, this is a quality impact socket set for automotive and industrial use.