Learn More
Grainger Thread Mill 5/8 -11 Carbide
Catalog No. 502767574
Encompass_Preferred
The Grainger 16R714 is a Westward 3/8-inch drive deep impact socket set. This 15-piece socket set includes sizes ranging from 4mm to 14mm, providing a wide variety of metric socket sizes for impact wrench applications. The sockets are made from chrome vanadium steel for durability and have a black phosphate finish to resist corrosion. The set comes organized in a sturdy plastic blow mold case for convenient storage and portability. With deep impact sockets suitable for most automotive and machinery applications, this Westward socket set is a versatile addition to any toolbox.
Catalog No. 50-276-7574 Supplier Grainger Supplier No. 16R714
The Grainger 16R714 is a Westward 3/8-inch drive deep impact socket set. This 15-piece socket set includes sizes ranging from 4mm to 14mm, providing a wide variety of metric socket sizes for impact wrench applications. The sockets are made from chrome vanadium steel for durability and have a black phosphate finish to resist corrosion. The set comes organized in a sturdy plastic blow mold case for convenient storage and portability. With deep impact sockets suitable for most automotive and machinery applications, this Westward socket set is a versatile addition to any toolbox.