Learn More
Grainger Thread Mill 9/16 -18 Carbide
Catalog No. 502765670
Encompass_Preferred
The Scientific Cutting Tools 16R712 thread mill is a solid carbide end mill designed for thread milling applications. It has an overall length of 3.5", a shank diameter of 0.5", and features a TiAlN coating for increased tool life and heat resistance. The thread mill has a 60 degree profile angle and can cut a variety of thread sizes and pitches. It is made from micro-grain carbide for strength and durability. This thread mill is well-suited for difficult to machine materials like stainless steel, titanium, and nickel alloys. It can be run at speeds up to 2000 SFM.
Catalog No. 50-276-5670 Supplier Grainger Supplier No. 16R712
The Scientific Cutting Tools 16R712 thread mill is a solid carbide end mill designed for thread milling applications. It has an overall length of 3.5", a shank diameter of 0.5", and features a TiAlN coating for increased tool life and heat resistance. The thread mill has a 60 degree profile angle and can cut a variety of thread sizes and pitches. It is made from micro-grain carbide for strength and durability. This thread mill is well-suited for difficult to machine materials like stainless steel, titanium, and nickel alloys. It can be run at speeds up to 2000 SFM.