Learn More
Grainger Thread Mill Carbide
Catalog No. 502766041
Encompass_Preferred
The Micro 100 16R744 is a 1/4" diameter, 4 flute, carbide thread mill with a TiAlN coating. It has a right hand spiral flute and can cut 60 degree threads. The overall length is 3" and it has a 1/2" shank. This thread mill is designed for high speed machining in steels, stainless steels, cast iron, and non-ferrous metals. It can be run at speeds up to 2000 SFM. The TiAlN coating provides heat and wear resistance. This is a high performance thread mill for precision threading operations.
Catalog No. 50-276-6041 Supplier Grainger Supplier No. 16R744
The Micro 100 16R744 is a 1/4" diameter, 4 flute, carbide thread mill with a TiAlN coating. It has a right hand spiral flute and can cut 60 degree threads. The overall length is 3" and it has a 1/2" shank. This thread mill is designed for high speed machining in steels, stainless steels, cast iron, and non-ferrous metals. It can be run at speeds up to 2000 SFM. The TiAlN coating provides heat and wear resistance. This is a high performance thread mill for precision threading operations.