Learn More
Grainger Thread Mill Carbide
Catalog No. 502764976
Encompass_Preferred
The Micro 100 16R765 is a 1/4" diameter, 3/4" long, 4 flute carbide thread mill with a right hand spiral flute. It has a TiAlN coating and can cut 60-62 HRC materials. The thread mill has a 1/4"-28 UNF shank and can cut thread sizes from #10-24 through 1/4"-28. It has a neck relief below the thread for improved chip evacuation. The Micro 100 16R765 thread mill is made in the USA.
Catalog No. 50-276-4976 Supplier Grainger Supplier No. 16R765
The Micro 100 16R765 is a 1/4" diameter, 3/4" long, 4 flute carbide thread mill with a right hand spiral flute. It has a TiAlN coating and can cut 60-62 HRC materials. The thread mill has a 1/4"-28 UNF shank and can cut thread sizes from #10-24 through 1/4"-28. It has a neck relief below the thread for improved chip evacuation. The Micro 100 16R765 thread mill is made in the USA.