Learn More
Grainger Thread Mill Carbide
Catalog No. 502767625
Encompass_Preferred
The Micro 100 16R769 is a carbide thread mill with a 1/4" shank diameter, 1/2" max cutting diameter, and 1/4"-28 internal thread size. It has a TiAlN coating for increased tool life and can machine a variety of materials including steel, stainless steel, cast iron, aluminum, and more. With its variable helix flutes, it provides efficient chip evacuation while milling. The 16R769 thread mill is made in the USA and well-suited for production threading applications.
Catalog No. 50-276-7625 Supplier Grainger Supplier No. 16R769
The Micro 100 16R769 is a carbide thread mill with a 1/4" shank diameter, 1/2" max cutting diameter, and 1/4"-28 internal thread size. It has a TiAlN coating for increased tool life and can machine a variety of materials including steel, stainless steel, cast iron, aluminum, and more. With its variable helix flutes, it provides efficient chip evacuation while milling. The 16R769 thread mill is made in the USA and well-suited for production threading applications.