Learn More
Grainger Thread Mill Carbide
Catalog No. 502768313
Encompass_Preferred
The Micro 100 16R770 is a carbide thread mill with a 1/2" shank diameter, 1-1/2" overall length, and 1/4"-28 internal thread size. It has a TiAlN coating for increased tool life and can machine a variety of materials including steel, stainless steel, cast iron, aluminum, and more. With its variable helix flutes, it provides efficient chip evacuation while threading, allowing for optimal performance. The 16R770 thread mill is made in the USA out of micrograin carbide for strength and durability. It's a high quality threading tool well-suited for production threading applications.
Catalog No. 50-276-8313 Supplier Grainger Supplier No. 16R770
The Micro 100 16R770 is a carbide thread mill with a 1/2" shank diameter, 1-1/2" overall length, and 1/4"-28 internal thread size. It has a TiAlN coating for increased tool life and can machine a variety of materials including steel, stainless steel, cast iron, aluminum, and more. With its variable helix flutes, it provides efficient chip evacuation while threading, allowing for optimal performance. The 16R770 thread mill is made in the USA out of micrograin carbide for strength and durability. It's a high quality threading tool well-suited for production threading applications.